Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176812.4 | NP_789782.1 | 3 | 600 | ||
GenBank mRNA | AB100261.1 | - | 3 | 545 | ||
AY329085.1 | - | 3 | 435 | |||
AY726594.1 | - | 3 | 447 | |||
BC033859.1 | - | 3 | 544 | 5173394 | ||
BQ926070.1 | - | 594 |