Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178468.5 | NP_848563.1 | 4 | 2303 | ||
GenBank mRNA | AK091116.1 | - | 5 | 1870 | ||
BC113483.1 | - | 4 | 2282 | 8322535 |