Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s43377
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
6
637
NM_001283116.2
NP_001270045.1
5
391
NM_001283129.2
NP_001270058.1
5
631
NM_001283148.2
NP_001270077.1
5
391
NM_001283154.2
NP_001270083.1
5
503
NM_001283199.2
NP_001270128.1
5
503
NM_001283215.2
NP_001270144.1
5
631
NM_001322141.1
NP_001309070.1
5
631
NM_001322142.1
NP_001309071.1
5
391
NM_001322143.1
NP_001309072.1
5
704
NM_001322144.1
NP_001309073.1
5
631
NM_001322146.1
NP_001309075.1
5
541
NM_001322148.1
NP_001309077.1
5
708
NM_001322149.1
NP_001309078.1
5
631
NM_001322160.1
NP_001309089.1
5
593
NM_152906.6
NP_690870.3
5
503
NR_136211.1
5
464
XM_011529863.1
XP_011528165.1
5
542
XM_011529865.1
XP_011528167.1
4
407
XM_017028577.1
XP_016884066.1
5
464
XM_017028578.1
XP_016884067.1
5
483
XM_017028579.1
XP_016884068.1
5
512
XM_017028580.1
XP_016884069.1
6
601
XM_017028581.1
XP_016884070.1
5
595
XM_017028582.1
XP_016884071.1
6
678
XM_017028583.1
XP_016884072.1
5
425
XM_017028584.1
XP_016884073.1
5
688
XM_017028587.1
XP_016884076.1
5
595
XM_017028588.1
XP_016884077.1
5
444
GenBank mRNA
AK289862.1
-
6
606
AK295210.1
-
4
517
AK297922.1
-
5
349
AK301366.1
-
5
590
AK316020.1
-
5
352
AK316056.1
-
5
611
AL713640.1
-
2
131
BC041339.1
-
5
466
5284126
BG685068.1
-
3
241
BI544307.1
-
5
466
BI911898.1
-
5
454
BM013784.1
-
5
675
BP303570.1
-
4
288
BU173566.1
-
5
423
CF552364.1
-
4
362
CR456355.1
-
4
318
CR627452.1
-
5
346
DB021425.1
-
3
181
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-77psw:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline