Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMID1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267895.1 | NP_001254824.1 | 14 | 1316 | ||
NM_133455.3 | NP_597712.2 | 14 | 1322 | |||
XM_011529869.2 | XP_011528171.1 | 15 | 1327 | |||
GenBank mRNA | AJ416090.1 | - | 14 | 1189 | ||
BC046358.1 | - | 14 | 1270 | 5770546 | ||
CR456407.1 | - | 14 | 1205 |