Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRABD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080824.2 | NP_001074293.1 | 5 | 1189 | ||
NM_001277053.1 | NP_001263982.1 | 6 | 1336 | |||
XM_011532504.1 | XP_011530806.1 | 6 | 1136 | |||
GenBank mRNA | AK128325.1 | - | 3 | 2228 | ||
AK296608.1 | - | 6 | 1173 | |||
BC051789.1 | - | 5 | 1190 | |||
JQ653415.1 | - | 6 | 1130 |