Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320417.1 | NP_001307346.1 | 4 | 618 | ||
NM_001320418.1 | NP_001307347.1 | 4 | 494 | |||
NM_138798.2 | NP_620153.1 | 4 | 494 | |||
XM_011510581.2 | XP_011508883.1 | 4 | 506 | |||
XM_011510582.2 | XP_011508884.1 | 4 | 506 | |||
XM_011510583.1 | XP_011508885.1 | 4 | 592 | |||
XM_017003315.1 | XP_016858804.1 | 4 | 506 | |||
GenBank mRNA | AL161992.1 | - | 2 | 2743 | ||
BC018453.1 | - | 4 | 462 | 4509694 | ||
BU507044.1 | - | 4 | 476 |