Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIS3L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257281.1 | NP_001244210.1 | 6 | 687 | ||
NM_001257282.1 | NP_001244211.1 | 6 | 687 | |||
NM_152383.4 | NP_689596.4 | 6 | 687 | |||
NR_046476.1 | 6 | 687 | ||||
NR_046477.1 | 6 | 687 | ||||
GenBank mRNA | AF443854.1 | - | 6 | 586 | ||
AK094293.1 | - | 6 | 688 | |||
AK295363.1 | - | 6 | 557 | |||
AL834174.1 | - | 4 | 226 | |||
BC026166.1 | - | 6 | 591 | 4822064 | ||
BC036113.1 | - | 6 | 586 | 5297794 | ||
BX648325.1 | - | 6 | 579 |