Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACMSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138326.2 | NP_612199.2 | 4 | 296 | ||
XM_005263589.4 | XP_005263646.1 | 5 | 704 | |||
GenBank mRNA | AB071418.1 | - | 4 | 229 | ||
AK125008.1 | - | 4 | 296 | |||
BC107420.1 | - | 4 | 236 | 4594027 |