Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACMSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307983.1 | NP_001294912.1 | 9 | 987 | ||
NM_138326.2 | NP_612199.2 | 9 | 930 | |||
XM_005263586.4 | XP_005263643.1 | 7 | 1080 | |||
XM_005263588.4 | XP_005263645.1 | 8 | 802 | |||
XM_005263589.4 | XP_005263646.1 | 10 | 1338 | |||
XM_005263590.4 | XP_005263647.1 | 6 | 1644 | |||
XM_011510592.2 | XP_011508894.1 | 9 | 1288 | |||
XM_017003325.1 | XP_016858814.1 | 9 | 1285 | |||
XM_017003326.1 | XP_016858815.1 | 9 | 1935 | |||
GenBank mRNA | AB071418.1 | - | 9 | 863 | ||
BC016018.1 | - | 9 | 989 | 4714682 | ||
BC107420.1 | - | 9 | 870 | 4594027 | ||
KU178859.1 | - | 6 | 689 | |||
KU178860.1 | - | 6 | 624 |