Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029881.2 | NP_001025052.1 | 5 | 452 | ||
XM_011532514.2 | XP_011530816.1 | 5 | 535 | |||
XM_011532517.2 | XP_011530819.1 | 3 | 822 | |||
XM_017003329.1 | XP_016858818.1 | 4 | 1306 | |||
XM_017003330.1 | XP_016858819.1 | 3 | 1216 | |||
GenBank mRNA | BC127684.1 | - | 5 | 452 | 40127392 | |
BC127685.1 | - | 5 | 450 | 40127393 | ||
BC140923.1 | - | 5 | 426 | 9056735 |