Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008387.2 | NP_001008388.1 | 6 | 685 | ||
NM_001270040.1 | NP_001256969.1 | 5 | 547 | |||
NM_198448.3 | NP_940850.1 | 6 | 585 | |||
XM_005264135.2 | XP_005264192.1 | 6 | 747 | |||
GenBank mRNA | AB161037.1 | - | 6 | 570 | ||
AB161038.1 | - | 6 | 670 | |||
AK292595.1 | - | 6 | 569 | |||
AY359047.1 | - | 6 | 571 | |||
AY428734.1 | - | 6 | 544 | |||
BC103852.2 | - | 5 | 506 | 40002928 | ||
BC103853.1 | - | 6 | 644 | 40002929 | ||
BC103854.1 | - | 6 | 544 | 40002931 | ||
CR541938.1 | - | 5 | 486 |