Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039569.1 | NP_001034658.1 | 2, 3 | 332 | ||
NR_110905.1 | 2, 3 | 332 | ||||
XM_011510600.2 | XP_011508902.1 | 2, 3 | 312 | |||
GenBank mRNA | AF393369.1 | - | 2, 3 | 214 | ||
AK299026.1 | - | 2, 3 | 312 | |||
BC009606.1 | - | 2, 3 | 333 | 4284350 | ||
BC021898.1 | - | 2, 3 | 276 | 4340257 | ||
BG177582.1 | - | 465 | ||||
BG340480.1 | - | 1, 2 | 94 |