Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DTNB SIRNA Assays ›
Gene Symbol:
DTNB
Gene Name:
dystrobrevin beta
Gene Aliases:
-
Chromosome Location:
Chr.2: 25377220 - 25673647 on Build GRCh38
UniGene ID:
Hs.307720
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s4349
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DTNB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256303.1
NP_001243232.1
4
428
NM_001256304.1
NP_001243233.1
3
165
NM_001256308.2
NP_001243237.1
3
299
NM_001320932.1
NP_001307861.1
4
428
NM_001320933.1
NP_001307862.1
4
428
NM_001320934.1
NP_001307863.1
4
428
NM_001320935.1
NP_001307864.1
4
428
NM_001320936.1
NP_001307865.1
4
428
NM_001320937.1
NP_001307866.1
4
428
NM_021907.4
NP_068707.1
4
428
NM_033147.3
NP_149159.2
4
428
NM_033148.3
NP_149160.1
4
428
NM_183360.2
NP_899204.1
4
428
NM_183361.2
NP_899205.1
4
428
XM_005264182.1
XP_005264239.1
4
345
XM_011532671.1
XP_011530973.1
4
345
XM_011532672.2
XP_011530974.1
4
345
XM_011532673.1
XP_011530975.1
4
345
XM_011532674.1
XP_011530976.1
4
345
XM_011532675.1
XP_011530977.1
4
345
XM_011532676.1
XP_011530978.1
4
345
XM_011532677.2
XP_011530979.1
3
801
XM_011532679.1
XP_011530981.1
4
345
XM_011532680.2
XP_011530982.1
4
352
XM_011532681.2
XP_011530983.1
4
345
XM_017003529.1
XP_016859018.1
4
345
XM_017003530.1
XP_016859019.1
4
345
XM_017003531.1
XP_016859020.1
4
345
XM_017003532.1
XP_016859021.1
4
345
XM_017003533.1
XP_016859022.1
4
345
XM_017003534.1
XP_016859023.1
4
345
XM_017003535.1
XP_016859024.1
4
345
XM_017003536.1
XP_016859025.1
4
345
XM_017003538.1
XP_016859027.1
4
277
XM_017003539.1
XP_016859028.1
4
288
XM_017003540.1
XP_016859029.1
4
345
XM_017003541.1
XP_016859030.1
4
329
XM_017003542.1
XP_016859031.1
4
352
XM_017003543.1
XP_016859032.1
4
332
XM_017003544.1
XP_016859033.1
4
352
XM_017003545.1
XP_016859034.1
4
332
GenBank mRNA
AF022728.1
-
4
326
AK094494.1
-
4
291
AK225724.1
-
4
331
AK294174.1
-
4
333
AK294357.1
-
4
195
AK300019.1
-
4
305
AK301386.1
-
3
301
AL555818.3
-
4
342
BC016655.1
-
4
321
3868207
BC049366.1
-
4
419
4817552
BT009805.1
-
3
165
BU846095.1
-
2
106
DB460644.1
-
4
428
DQ160290.1
-
3
165
Y12712.1
-
4
348
Y15718.1
-
3
165
Y15722.1
-
3
165
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-8skhh:80/10.239.164.194:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline