Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317002.1 | NP_001303931.1 | 5, 6 | 456 | ||
NM_001317003.1 | NP_001303932.1 | 3, 4 | 566 | |||
NM_001317004.1 | NP_001303933.1 | 4, 5 | 367 | |||
NM_001317005.1 | NP_001303934.1 | 4, 5 | 627 | |||
NM_001317006.1 | NP_001303935.1 | 2, 3 | 495 | |||
NM_177964.4 | NP_808879.2 | 4, 5 | 638 | |||
XM_005246306.4 | XP_005246363.1 | 6, 7 | 608 | |||
XM_006712279.2 | XP_006712342.1 | 4, 5 | 460 | |||
XM_006712280.3 | XP_006712343.1 | 6, 7 | 606 | |||
XM_006712281.3 | XP_006712344.1 | 4, 5 | 380 | |||
XM_011510620.2 | XP_011508922.1 | 6, 7 | 481 | |||
XM_011510623.2 | XP_011508925.1 | 5, 6 | 454 | |||
XM_011510625.1 | XP_011508927.1 | 7, 8 | 685 | |||
XM_017003364.1 | XP_016858853.1 | 5, 6 | 466 | |||
XM_017003365.1 | XP_016858854.1 | 4, 5 | 393 | |||
XM_017003366.1 | XP_016858855.1 | 4, 5 | 437 | |||
GenBank mRNA | AF435957.1 | - | 4, 5 | 367 | ||
BC018203.1 | - | 3, 4 | 315 | 3861272 | ||
BC040176.2 | - | 4, 5 | 638 | 4794338 | ||
BQ056975.1 | - | 2, 3 | 232 | |||
CN359800.1 | - | 4, 5 | 327 | |||
DB461489.1 | - | 5, 6 | 449 |