Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HBEGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001945.2 | NP_001936.1 | 4 | 736 | ||
GenBank mRNA | AK222688.1 | - | 4 | 736 | ||
AK307310.1 | - | 3 | 457 | |||
AK313202.1 | - | 4 | 737 | |||
BC033097.1 | - | 4 | 723 | 5424240 | ||
M60278.1 | - | 4 | 722 |