Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182500.2 | NP_872306.1 | 2 | 567 | ||
XM_006711857.2 | XP_006711920.1 | 2 | 491 | |||
XM_006711858.2 | XP_006711921.1 | 2 | 491 | |||
XM_006711859.3 | XP_006711922.1 | 2 | 491 | |||
GenBank mRNA | AK057872.1 | - | 2 | 573 | ||
AK292828.1 | - | 2 | 492 | |||
BC120930.2 | - | 2 | 534 | 40114857 |