Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174899.4 | NP_777559.3 | 2 | 160 | ||
XM_005246317.1 | XP_005246374.1 | 2 | 170 | |||
GenBank mRNA | AK123085.1 | - | 3 | 465 | ||
AK289655.1 | - | 2 | 145 | |||
AK309702.1 | - | 2 | 138 | |||
BC022538.1 | - | 3 | 276 | 4817261 | ||
BC033935.1 | - | 2 | 153 | 5296808 | ||
BF674223.1 | - | 2 | 160 |