Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTYMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165031.1 | NP_001158503.1 | 2, 3 | 432 | ||
NM_012145.3 | NP_036277.2 | 2, 3 | 432 | |||
GenBank mRNA | AK223434.1 | - | 2, 3 | 279 | ||
BC001827.2 | - | 2, 3 | 272 | 3533327 | ||
BC147006.1 | - | 2, 3 | 371 | 9056934 | ||
BC147010.1 | - | 2, 3 | 371 | 9056938 | ||
BC171894.1 | - | 2, 3 | 367 | |||
BC171902.1 | - | 2, 3 | 371 | 9054556 | ||
BM552707.1 | - | 2, 3 | 418 | |||
CR541986.1 | - | 2, 3 | 247 | |||
CR542015.1 | - | 2, 3 | 247 | |||
CX166352.1 | - | 2, 3 | 432 | |||
L16991.1 | - | 2, 3 | 289 | |||
X54729.1 | - | 2, 3 | 271 |