Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144633.2 | NP_653234.2 | 7 | 1314 | ||
XM_011533347.2 | XP_011531649.1 | 3 | 473 | |||
XM_017005698.1 | XP_016861187.1 | 7 | 1385 | |||
XM_017005699.1 | XP_016861188.1 | 6 | 1051 | |||
XM_017005701.1 | XP_016861190.1 | 8 | 1455 | |||
XM_017005702.1 | XP_016861191.1 | 6 | 1051 | |||
GenBank mRNA | AB209053.1 | - | 8 | 1388 | ||
AK294759.1 | - | 2 | 264 | |||
AK295633.1 | - | 7 | 1361 | |||
AY053503.1 | - | 7 | 1314 |