Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190233.1 | NP_001177162.1 | 5, 6 | 456 | ||
NM_145261.3 | NP_660304.1 | 5, 6 | 444 | |||
NR_033721.1 | 4, 5 | 392 | ||||
NR_033722.1 | 4, 5 | 364 | ||||
NR_033723.1 | 5, 6 | 389 | ||||
NR_046073.1 | 2, 3 | 238 | ||||
GenBank mRNA | AK311765.1 | - | 5, 6 | 372 | ||
BC009702.1 | - | 5, 6 | 396 | 3892297 | ||
BC073989.1 | - | 5, 6 | 389 | 5760865 | ||
BE773017.1 | - | 5, 6 | 411 | |||
BF666599.1 | - | 5, 6 | 406 | |||
BG492744.1 | - | 4, 5 | 329 | |||
BM701756.1 | - | 4, 5 | 317 | |||
BM702203.1 | - | 4, 5 | 317 | |||
BQ438379.1 | - | 2, 3 | 175 | |||
CB052107.1 | - | 5, 6 | 327 | |||
CB054787.1 | - | 2, 3 | 160 | |||
CB159371.1 | - | 5, 6 | 456 | |||
KU178861.1 | - | 4, 5 | 272 | |||
KU178862.1 | - | 3, 4 | 192 |