Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190233.1 | NP_001177162.1 | 5 | 439 | ||
NM_145261.3 | NP_660304.1 | 5 | 427 | |||
NR_033721.1 | 4 | 375 | ||||
NR_033722.1 | 4 | 347 | ||||
NR_033723.1 | 5 | 372 | ||||
NR_046073.1 | 2 | 221 | ||||
GenBank mRNA | AI268262.1 | - | 5 | 426 | ||
AK311765.1 | - | 5 | 355 | |||
BC009702.1 | - | 5 | 379 | 3892297 | ||
BC073989.1 | - | 5 | 372 | 5760865 | ||
BE773017.1 | - | 5 | 394 | |||
BF666599.1 | - | 5 | 389 | |||
BG492744.1 | - | 4 | 312 | |||
BM701756.1 | - | 4 | 300 | |||
BM702203.1 | - | 4 | 300 | |||
BQ438379.1 | - | 2 | 158 | |||
CB052107.1 | - | 5 | 310 | |||
CB054787.1 | - | 2 | 143 | |||
CB159371.1 | - | 5 | 439 | |||
KU178861.1 | - | 4 | 255 | |||
KU178862.1 | - | 3 | 175 |