Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153605.3 | NP_705833.3 | 19 | 8607 | ||
XM_005247117.4 | XP_005247174.1 | 18 | 8310 | |||
GenBank mRNA | AF451988.1 | - | 7 | 724 | ||
AK091199.1 | - | 11 | 1078 | |||
AK098569.1 | - | 3 | 399 | |||
AL831927.1 | - | 7 | 630 | |||
BC036247.1 | - | 2 | 150 | |||
BC050394.1 | - | 4 | 390 | |||
BX647079.1 | - | 11 | 1095 | |||
BX648353.1 | - | 16 | 2657 | |||
CR749313.1 | - | 16 | 1732 |