Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102608.1 | NP_001096078.1 | 5, 6 | 2434 | ||
XM_005247121.4 | XP_005247178.1 | 7, 8 | 2593 | |||
XM_011512423.2 | XP_011510725.1 | 6, 7 | 2658 | |||
XM_011512424.2 | XP_011510726.1 | 6, 7 | 2658 | |||
XM_011512425.2 | XP_011510727.1 | 8, 9 | 2773 | |||
XM_011512426.2 | XP_011510728.1 | 8, 9 | 2667 | |||
XM_011512428.2 | XP_011510730.1 | 7, 8 | 2694 | |||
XM_011512431.2 | XP_011510733.1 | 6, 7 | 2659 | |||
XM_011512432.2 | XP_011510734.1 | 6, 7 | 2659 | |||
XM_017005711.1 | XP_016861200.1 | 9, 10 | 2878 | |||
XM_017005712.1 | XP_016861201.1 | 8, 9 | 2982 | |||
XM_017005713.1 | XP_016861202.1 | 9, 10 | 2929 | |||
XM_017005714.1 | XP_016861203.1 | 7, 8 | 9034 | |||
XM_017005715.1 | XP_016861204.1 | 8, 9 | 2799 | |||
XM_017005716.1 | XP_016861205.1 | 6, 7 | 2659 | |||
GenBank mRNA | AM774225.1 | - | 5, 6 | 2434 | ||
AM774226.1 | - | 1, 2 | 134 |