Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301790.1 | NP_001288719.1 | 5 | 880 | ||
NM_001301792.1 | NP_001288721.1 | 5 | 901 | |||
NM_152396.3 | NP_689609.2 | 6 | 1015 | |||
XM_005264867.3 | XP_005264924.1 | 6 | 1015 | |||
XM_006712970.3 | XP_006713033.1 | 6 | 891 | |||
XM_011533357.2 | XP_011531659.1 | 6 | 959 | |||
XM_017005718.1 | XP_016861207.1 | 6 | 891 | |||
XM_017005719.1 | XP_016861208.1 | 5 | 873 | |||
XM_017005722.1 | XP_016861211.1 | 5 | 873 | |||
GenBank mRNA | AK027185.1 | - | 2 | 128 | ||
AK293370.1 | - | 5 | 873 | |||
BC022400.1 | - | 6 | 972 | 4693314 | ||
CX872865.1 | - | 3 | 384 |