Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17RE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193380.1 | NP_001180309.1 | 6 | 677 | ||
NM_153480.1 | NP_705613.1 | 6 | 677 | |||
NM_153481.1 | NP_705614.1 | 4 | 541 | |||
NM_153483.2 | NP_705616.2 | 7 | 696 | |||
NR_104198.1 | 5 | 661 | ||||
XM_006712975.1 | XP_006713038.1 | 5 | 503 | |||
XM_006712976.1 | XP_006713039.1 | 5 | 641 | |||
XM_011533361.2 | XP_011531663.1 | 6 | 593 | |||
XM_011533362.2 | XP_011531664.1 | 6 | 612 | |||
XM_011533363.2 | XP_011531665.1 | 6 | 612 | |||
XM_011533364.2 | XP_011531666.1 | 6 | 612 | |||
GenBank mRNA | AF458069.1 | - | 6 | 677 | ||
AK074238.1 | - | 6 | 657 | |||
AK096793.1 | - | 5 | 639 | |||
AK096801.1 | - | 4 | 521 | |||
AK297697.1 | - | 6 | 643 | |||
AK298025.1 | - | 3 | 423 | |||
AK314476.1 | - | 6 | 662 | |||
AY358451.1 | - | 6 | 677 | |||
BC035478.1 | - | 7 | 689 | |||
BC063110.1 | - | 6 | 675 | |||
BC137077.1 | - | 7 | 639 | 9021074 | ||
BC137078.1 | - | 7 | 639 | 9021075 |