Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145262.3 | NP_660305.2 | 5 | 1621 | ||
NR_026699.1 | 5 | 1623 | ||||
NR_026700.1 | 5 | 729 | ||||
NR_026701.1 | 5 | 1621 | ||||
NR_026702.1 | 4 | 659 | ||||
XM_017005730.1 | XP_016861219.1 | 4 | 1467 | |||
GenBank mRNA | AF448855.1 | - | 5 | 1622 | ||
AK074215.1 | - | 5 | 714 | |||
AY134474.1 | - | 3 | 1327 | |||
AY172690.1 | - | 4 | 659 | |||
BC021896.1 | - | 4 | 1383 | 3917295 | ||
BC036862.1 | - | 5 | 1623 | |||
BC042151.1 | - | 5 | 1585 | 4577225 | ||
DQ352863.1 | - | 5 | 1049 |