Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145262.3 | NP_660305.2 | 5 | 1557 | ||
NR_026699.1 | 5 | 1559 | ||||
NR_026701.1 | 5 | 1557 | ||||
XM_017005730.1 | XP_016861219.1 | 4 | 1403 | |||
GenBank mRNA | AF448855.1 | - | 5 | 1558 | ||
AY134474.1 | - | 3 | 1263 | |||
BC021896.1 | - | 4 | 1319 | 3917295 | ||
BC036862.1 | - | 5 | 1559 | |||
BC042151.1 | - | 5 | 1521 | 4577225 |