Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317837.1 | NP_001304766.1 | 5 | 499 | ||
NM_001317838.1 | NP_001304767.1 | 5 | 496 | |||
NM_001317839.1 | NP_001304768.1 | 5 | 374 | |||
NM_001317841.1 | NP_001304770.1 | 4 | 789 | |||
NM_001317842.1 | NP_001304771.1 | 4 | 633 | |||
NM_152399.3 | NP_689612.3 | 5 | 655 | |||
XM_005262737.2 | XP_005262794.1 | 5 | 463 | |||
XM_017007725.1 | XP_016863214.1 | 5 | 369 | |||
XM_017007726.1 | XP_016863215.1 | 4 | 309 | |||
GenBank mRNA | AK055396.1 | - | 5 | 376 | ||
AK310583.1 | - | 4 | 508 | |||
AL552582.3 | - | 5 | 526 | |||
BC047657.1 | - | 5 | 655 | 5264721 | ||
BX359599.2 | - | 4 | 661 | |||
DA176165.1 | - | 5 | 460 | |||
DB116307.1 | - | 4 | 508 |