Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114734.1 | NP_001108206.2 | 2 | 773 | ||
XM_017007727.1 | XP_016863216.1 | 2 | 812 |