Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304358.1 | NP_001291287.1 | 3 | 929 | ||
NM_174900.4 | NP_777560.2 | 4 | 1095 | |||
XM_011531604.2 | XP_011529906.1 | 3 | 950 | |||
XM_011531605.1 | XP_011529907.1 | 4 | 1100 | |||
XM_011531606.1 | XP_011529908.1 | 4 | 1104 | |||
XM_011531607.2 | XP_011529909.1 | 3 | 947 | |||
GenBank mRNA | AF450454.1 | - | 1 | 687 | ||
AK056719.1 | - | 3 | 929 | |||
AY429593.1 | - | 4 | 1095 | |||
CN420210.1 | - | 1 | 29 |