Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115007.2 | NP_001108479.1 | 6 | 595 | ||
NM_001115008.2 | NP_001108480.1 | 6 | 796 | |||
NM_001288996.1 | NP_001275925.1 | 3 | 1192 | |||
NM_001288997.1 | NP_001275926.1 | 6 | 533 | |||
NM_194282.3 | NP_919258.2 | 6 | 1459 | |||
NR_110254.1 | 5 | 792 | ||||
XM_005262750.4 | XP_005262807.1 | 5 | 1268 | |||
XM_006714081.3 | XP_006714144.1 | 5 | 1194 | |||
XM_017007728.1 | XP_016863217.1 | 3 | 1001 | |||
GenBank mRNA | AB111889.1 | - | 5 | 779 | ||
AK292769.1 | - | 6 | 543 | |||
AK294256.1 | - | 6 | 1388 | |||
BC109278.2 | - | 3 | 1067 | 40028371 | ||
BX537919.1 | - | 5 | 693 | |||
BX640657.1 | - | 6 | 522 | |||
BX640966.1 | - | 6 | 1454 |