Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNPDA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270880.1 | NP_001257809.1 | 3 | 393 | ||
NM_001270881.1 | NP_001257810.1 | 3 | 336 | |||
NM_138335.2 | NP_612208.1 | 4 | 495 | |||
NR_073094.1 | 3 | 336 | ||||
GenBank mRNA | AB209904.1 | - | 4 | 406 | ||
AF247786.1 | - | 4 | 456 | |||
AK055639.1 | - | 4 | 415 | |||
AK296051.1 | - | 3 | 225 | |||
AY173948.1 | - | 3 | 330 | |||
BC015532.2 | - | 3 | 326 | 3914747 | ||
BP274548.1 | - | 4, 5 | 495 | |||
HY180477.1 | - | 3 | 239 |