Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300754.1 | NP_001287683.1 | 9 | 1271 | ||
NM_001300756.1 | NP_001287685.1 | 9 | 1637 | |||
NM_178833.5 | NP_849155.2 | 10 | 1808 | |||
XM_006714085.2 | XP_006714148.1 | 10 | 1807 | |||
XM_006714086.3 | XP_006714149.1 | 9 | 1636 | |||
GenBank mRNA | AK172823.1 | - | 10 | 1808 | ||
AK297010.1 | - | 9 | 1182 | |||
AK299888.1 | - | 9 | 1196 | |||
BC009732.1 | - | 7 | 876 | |||
BC047447.1 | - | 10 | 1338 | 5295722 |