Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL31RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242636.1 | NP_001229565.1 | 8 | 1129 | ||
NM_001242637.1 | NP_001229566.1 | 8 | 1138 | |||
NM_001242638.1 | NP_001229567.1 | 10 | 1502 | |||
NM_001242639.1 | NP_001229568.1 | 9 | 1273 | |||
NM_001297570.1 | NP_001284499.1 | 8 | 1138 | |||
NM_001297572.1 | NP_001284501.1 | 8 | 1266 | |||
NM_139017.5 | NP_620586.3 | 8 | 1138 | |||
XM_011543142.2 | XP_011541444.1 | 8 | 971 | |||
XM_011543143.2 | XP_011541445.1 | 8 | 1161 | |||
XM_011543144.2 | XP_011541446.1 | 7 | 1043 | |||
XM_011543145.2 | XP_011541447.1 | 5 | 612 | |||
GenBank mRNA | AF106913.1 | - | 8 | 1002 | ||
AF486620.1 | - | 7 | 850 | |||
AK297938.1 | - | 8 | 1250 | |||
AY358117.1 | - | 7 | 850 | |||
AY358740.1 | - | 8 | 1138 | |||
AY499339.1 | - | 8 | 1011 | |||
AY499340.1 | - | 8 | 1011 | |||
AY499341.1 | - | 8 | 1011 | |||
AY499342.1 | - | 10 | 1385 | |||
BC110490.2 | - | 9 | 1065 | 40007489 |