Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198449.2 | NP_940851.1 | 3 | 577 | ||
XM_011543146.2 | XP_011541448.1 | 4 | 359 | |||
GenBank mRNA | AK300860.1 | - | 3 | 542 | ||
AK304226.1 | - | 3 | 448 | |||
BC059398.1 | - | 3 | 544 | 30332712 |