Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198449.2 | NP_940851.1 | 7, 8 | 1157 | ||
XM_011543146.2 | XP_011541448.1 | 8, 9 | 939 | |||
GenBank mRNA | AK300860.1 | - | 7, 8 | 1122 | ||
AK304226.1 | - | 7, 8 | 1028 | |||
BC059398.1 | - | 7, 8 | 1124 | 30332712 |
Gene Symbol: EMBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003955.1 | 6, 7 | 1060 | |||
GenBank mRNA | AK128714.1 | - | 6, 7 | 1060 |