Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLCO6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289002.1 | NP_001275931.1 | 8 | 1620 | ||
NM_001289004.1 | NP_001275933.1 | 7 | 1434 | |||
NM_173488.4 | NP_775759.3 | 8 | 1620 | |||
XM_005271874.3 | XP_005271931.1 | 8 | 4817 | |||
XM_011543147.2 | XP_011541449.1 | 8 | 4712 | |||
XM_011543148.2 | XP_011541450.1 | 8 | 4580 | |||
XM_011543150.1 | XP_011541452.1 | 7 | 830 | |||
XM_011543153.2 | XP_011541455.1 | 6 | 710 | |||
GenBank mRNA | AF505657.1 | - | 8 | 1591 | ||
AY273897.1 | - | 8 | 1583 | |||
BC034976.2 | - | 7 | 1436 | 4820512 |