Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPARGC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172698.1 | NP_001166169.1 | 7 | 1854 | ||
NM_001172699.1 | NP_001166170.1 | 7 | 1885 | |||
NM_133263.3 | NP_573570.3 | 8 | 1971 | |||
XM_005268372.4 | XP_005268429.1 | 8 | 2165 | |||
XM_011537553.2 | XP_011535855.1 | 8 | 1921 | |||
XM_011537554.2 | XP_011535856.1 | 8 | 2168 | |||
XM_011537555.2 | XP_011535857.1 | 7 | 1796 | |||
XM_011537556.2 | XP_011535858.1 | 7 | 3144 | |||
XM_011537557.1 | XP_011535859.1 | 7 | 1884 | |||
GenBank mRNA | AF468496.1 | - | 8 | 1925 | ||
AF468497.1 | - | 7 | 1808 | |||
AK095391.1 | - | 8 | 1912 | |||
AK123614.1 | - | 7 | 1885 | |||
AY188947.1 | - | 8 | 1922 | |||
AY188948.1 | - | 8 | 1922 | |||
AY188949.1 | - | 8 | 2097 | |||
AY188950.1 | - | 8 | 2097 | |||
BC026956.1 | - | 4 | 939 | |||
BC132971.1 | - | 8 | 1971 | 40146993 | ||
BC132973.1 | - | 8 | 1971 | 40146995 | ||
BC144251.1 | - | 7 | 1854 | 9052771 |