Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MROH2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173489.4 | NP_775760.3 | 4 | 797 | ||
XM_011513952.1 | XP_011512254.1 | 4 | 783 | |||
XM_011513953.1 | XP_011512255.1 | 3 | 721 | |||
GenBank mRNA | BC034600.1 | - | 4 | 761 | ||
BC052962.1 | - | 4 | 758 | 4838071 |