Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286808.1 | NP_001273737.1 | 3 | 447 | ||
NM_130809.4 | NP_570721.1 | 3 | 447 | |||
XM_006714529.3 | XP_006714592.1 | 3 | 655 | |||
GenBank mRNA | AJ427339.1 | - | 3 | 400 | ||
AJ441110.3 | - | 3 | 400 | |||
AJ496322.1 | - | 3 | 400 | |||
AJ515158.1 | - | 3 | 400 | |||
AJ515428.1 | - | 3 | 400 | |||
AJ515429.1 | - | 3 | 400 | |||
AK057437.1 | - | 3 | 384 | |||
AK090486.1 | - | 2 | 273 | |||
AL831963.1 | - | 3 | 409 | |||
AL832056.1 | - | 3 | 410 | |||
AL832600.2 | - | 3 | 406 | |||
BC017066.1 | - | 3 | 384 | 3913311 | ||
DA605419.1 | - | 3 | 439 | |||
DA808343.1 | - | 3 | 447 |