Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152404.3 | NP_689617.3 | 6 | 1457 | ||
XM_005248243.4 | XP_005248300.1 | 6 | 1413 | |||
XM_011513957.2 | XP_011512259.1 | 6 | 1186 | |||
XM_011513958.2 | XP_011512260.1 | 5 | 1543 | |||
XM_011513959.2 | XP_011512261.1 | 7 | 1349 | |||
XM_011513961.2 | XP_011512263.1 | 5 | 1106 | |||
GenBank mRNA | AK057066.1 | - | 6 | 1421 | ||
AK091977.1 | - | 3 | 440 | |||
AK125803.1 | - | 7 | 2774 | |||
AK290809.1 | - | 6 | 1414 | |||
AK296136.1 | - | 5 | 1355 | |||
AK303770.1 | - | 6 | 1421 | |||
AK308046.1 | - | 6 | 1364 | |||
AK315851.1 | - | 6 | 1421 | |||
BC068446.1 | - | 6 | 1324 | 5295502 |