Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152405.4 | NP_689618.4 | 4 | 2000 | ||
XM_005248430.2 | XP_005248487.1 | 4 | 2000 | |||
XM_011543155.2 | XP_011541457.1 | 4 | 2000 | |||
GenBank mRNA | AK095189.1 | - | 3 | 401 | ||
BC130624.1 | - | 3 | 426 | 40146655 | ||
CN372786.1 | - | 4 | 640 |