Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102609.2 | NP_001096079.1 | 4 | 375 | ||
NM_001305393.1 | NP_001292322.1 | 4 | 394 | |||
NM_001305394.1 | NP_001292323.1 | 4 | 383 | |||
NM_001305395.1 | NP_001292324.1 | 5 | 549 | |||
NM_001305396.1 | NP_001292325.1 | 5 | 538 | |||
NM_001305397.1 | NP_001292326.1 | 5 | 522 | |||
XM_011534432.2 | XP_011532734.1 | 4 | 846 | |||
XM_017009029.1 | XP_016864518.1 | 5 | 1736 | |||
XM_017009030.1 | XP_016864519.1 | 5 | 1708 | |||
XM_017009031.1 | XP_016864520.1 | 4 | 435 | |||
XM_017009032.1 | XP_016864521.1 | 4 | 359 | |||
XM_017009033.1 | XP_016864522.1 | 4 | 426 | |||
GenBank mRNA | BC092511.1 | - | 3 | 1355 | ||
BU561730.1 | - | 5 | 485 | |||
BX100176.1 | - | 3 | 295 | |||
HY000873.1 | - | 4 | 461 | |||
HY001392.1 | - | 4 | 445 | |||
HY005987.1 | - | 4 | 394 | |||
HY016595.1 | - | 4 | 300 | |||
HY038777.1 | - | 4 | 375 |