Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012339.2 | NP_001012339.2 | 3 | 489 | ||
NM_194283.3 | NP_919259.3 | 3 | 489 | |||
XM_005248249.4 | XP_005248306.2 | 3 | 542 | |||
XM_005248250.3 | XP_005248307.2 | 3 | 542 | |||
XM_011513965.2 | XP_011512267.2 | 3 | 542 | |||
XM_011513966.2 | XP_011512268.2 | 3 | 542 | |||
GenBank mRNA | AY177688.1 | - | 3 | 498 | ||
AY258422.1 | - | 3 | 492 | |||
BC065745.1 | - | 3 | 499 |