Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRPEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152407.3 | NP_689620.2 | 3, 4 | 448 | ||
GenBank mRNA | AK074293.1 | - | 3, 4 | 334 | ||
AL832325.1 | - | 3, 4 | 343 | |||
BC036678.1 | - | 3, 4 | 368 | 5259464 | ||
BC070090.1 | - | 3, 4 | 350 | 30347705 |