Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099271.1 | NP_001092741.1 | 6 | 724 | ||
NM_152408.2 | NP_689621.2 | 5 | 523 | |||
XM_005248436.1 | XP_005248493.1 | 6 | 724 | |||
XM_011543158.2 | XP_011541460.1 | 6 | 724 | |||
XM_011543159.1 | XP_011541461.1 | 7 | 730 | |||
XM_011543160.1 | XP_011541462.1 | 5 | 584 | |||
XM_017009037.1 | XP_016864526.1 | 6 | 724 | |||
XM_017009038.1 | XP_016864527.1 | 5 | 644 | |||
XM_017009039.1 | XP_016864528.1 | 5 | 563 | |||
XM_017009040.1 | XP_016864529.1 | 5 | 527 | |||
GenBank mRNA | AK093098.1 | - | 5 | 493 | ||
AK296071.1 | - | 6 | 641 | |||
BC065750.1 | - | 6 | 670 | |||
BC101325.1 | - | 5 | 489 | 40024984 | ||
BC101326.2 | - | 5 | 493 | 40024987 | ||
BC101327.2 | - | 5 | 493 | 40024989 | ||
BC101328.2 | - | 5 | 489 |