Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099271.1 | NP_001092741.1 | 10 | 1424 | ||
NM_152408.2 | NP_689621.2 | 9 | 1223 | |||
XM_005248436.1 | XP_005248493.1 | 10 | 1340 | |||
XM_011543158.2 | XP_011541460.1 | 10 | 1424 | |||
XM_011543159.1 | XP_011541461.1 | 11 | 1430 | |||
XM_011543160.1 | XP_011541462.1 | 9 | 1284 | |||
XM_017009037.1 | XP_016864526.1 | 9 | 1319 | |||
XM_017009038.1 | XP_016864527.1 | 9 | 1344 | |||
XM_017009039.1 | XP_016864528.1 | 9 | 1263 | |||
XM_017009040.1 | XP_016864529.1 | 8 | 1122 | |||
GenBank mRNA | AK093098.1 | - | 9 | 1193 | ||
AK296071.1 | - | 10 | 1341 | |||
BC101325.1 | - | 9 | 1189 | 40024984 | ||
BC101326.2 | - | 9 | 1193 | 40024987 | ||
BC101327.2 | - | 9 | 1193 | 40024989 | ||
BC101328.2 | - | 9 | 1190 |