Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194251.2 | NP_919227.2 | 1 | 970 | ||
GenBank mRNA | AY351676.1 | - | 1 | 970 |
Set Membership: |
G-protein coupled receptor |