Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135586.1 | NP_001129058.1 | 2 | 543 | ||
NM_001300894.1 | NP_001287823.1 | 2 | 543 | |||
NM_152409.3 | NP_689622.2 | 2 | 356 | |||
XM_005271889.3 | XP_005271946.1 | 2 | 1192 | |||
XM_017009049.1 | XP_016864538.1 | 2 | 711 | |||
XM_017009050.1 | XP_016864539.1 | 2 | 319 | |||
GenBank mRNA | AJ437658.2 | - | 2 | 415 | ||
AK094881.1 | - | 2 | 493 | |||
BC053677.1 | - | 2 | 356 | 6166677 | ||
BX648529.1 | - | 2 | 423 | |||
DA433671.1 | - | 2 | 543 | |||
DA760705.1 | - | 2 | 545 |