Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLVS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010852.3 | NP_001010852.2 | 6 | 2255 | ||
XM_017010266.1 | XP_016865755.1 | 6 | 3957 | |||
GenBank mRNA | AK095527.1 | - | 6 | 2259 | ||
AK295158.1 | - | 6 | 717 | |||
BC101376.1 | - | 6 | 1789 | 40025886 | ||
BC101377.2 | - | 6 | 1791 | 40025889 |